siRNA Details
| Gene Symbol: Rgs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019344.2 | NP_062217.1 | 7 | 876 | ||
| XM_006250015.2 | XP_006250077.1 | 6 | 747 | |||
| XM_006250016.2 | XP_006250078.1 | 7 | 691 | |||
| XM_006250017.2 | XP_006250079.1 | 7 | 1041 | |||
| XM_008769675.1 | XP_008767897.1 | 7 | 1042 | |||
| XM_008769676.1 | XP_008767898.1 | 7 | 1207 | |||
| GenBank mRNA | AB006013.1 | - | 6 | 785 | ||
| BC089064.1 | - | 7 | 876 | 7317138 | ||

Rgs8
Genomic Map