siRNA Details
| Gene Symbol: Ncor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271103.1 | NP_001258032.1 | 39 | 6415 | ||
| GenBank mRNA | AF124821.1 | - | 4 | 543 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncor1| Gene Symbol: Ncor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271103.1 | NP_001258032.1 | 39 | 6415 | ||
| GenBank mRNA | AF124821.1 | - | 4 | 543 | ||