siRNA Details
Gene Symbol: Slc12a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019345.3 | NP_062218.3 | 1 | 176 | ||
XM_008772323.1 | XP_008770545.1 | 1 | 209 | |||
XM_008772324.1 | XP_008770546.1 | 1 | 209 | |||
XM_008772325.1 | XP_008770547.1 | 1 | 209 | |||
XM_008772326.1 | XP_008770548.1 | 1 | 209 | |||
XM_008772327.1 | XP_008770549.1 | 1 | 209 | |||
XM_008772328.1 | XP_008770550.1 | 1 | 209 | |||
GenBank mRNA | U10097.2 | - | 1 | 176 |
