siRNA Details
| Gene Symbol: SNTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006750.3 | NP_006741.1 | 3 | 996 | ||
| GenBank mRNA | BC048215.1 | - | 3 | 981 | 5805707 | |
| U40572.1 | - | 3 | 996 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNTB2| Gene Symbol: SNTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006750.3 | NP_006741.1 | 3 | 996 | ||
| GenBank mRNA | BC048215.1 | - | 3 | 981 | 5805707 | |
| U40572.1 | - | 3 | 996 | |||