siRNA Details
| Gene Symbol: Ucp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019354.3 | NP_062227.2 | 6 | 981 | ||
| GenBank mRNA | AB005143.1 | - | 4 | 608 | ||
| AB006613.1 | - | 4 | 608 | |||
| AB010743.1 | - | 6 | 952 | |||
| AF039033.1 | - | 4 | 608 | |||
| BC062230.1 | - | 6 | 953 | 6920447 | ||
| FQ227097.1 | - | 6 | 982 | |||
| FQ230356.1 | - | 2 | 267 | |||

Ucp2
Genomic Map