siRNA Details
| Gene Symbol: Cnn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019359.1 | NP_062232.1 | 7 | 1103 | ||
| GenBank mRNA | BC062020.1 | - | 7 | 1106 | 5623281 | |
| FQ209647.1 | - | 2 | 326 | |||
| FQ228936.1 | - | 4 | 540 | |||
| U06755.1 | - | 7 | 1103 | |||

Cnn3
Genomic Map