siRNA Details
| Gene Symbol: Rassf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019365.3 | NP_062238.1 | 1 | 435 | ||
| GenBank mRNA | AF002251.1 | - | 1 | 435 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rassf5| Gene Symbol: Rassf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019365.3 | NP_062238.1 | 1 | 435 | ||
| GenBank mRNA | AF002251.1 | - | 1 | 435 | ||