siRNA Details
| Gene Symbol: Crlf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168612.1 | NP_001162083.1 | 3 | 507 | ||
| GenBank mRNA | AF072835.1 | - | 415 | |||
| CB611803.1 | - | 508 | ||||
| CO555844.1 | - | 3 | 506 | |||
| DY321186.1 | - | 3 | 500 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crlf3| Gene Symbol: Crlf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168612.1 | NP_001162083.1 | 3 | 507 | ||
| GenBank mRNA | AF072835.1 | - | 415 | |||
| CB611803.1 | - | 508 | ||||
| CO555844.1 | - | 3 | 506 | |||
| DY321186.1 | - | 3 | 500 | |||