siRNA Details
Gene Symbol: Crlf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168612.1 | NP_001162083.1 | 6 | 1169 | ||
GenBank mRNA | AF072835.1 | - | 1077 | |||
CV075881.1 | - | 419 | ||||
DY560460.1 | - | 2 | 268 |
