siRNA Details
| Gene Symbol: Bet1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019368.3 | NP_062241.2 | 3 | 193 | ||
| XM_006230574.2 | XP_006230636.1 | 4 | 405 | |||
| GenBank mRNA | AF003998.1 | - | 3 | 232 | ||
| BC059138.1 | - | 3 | 197 | 6890305 | ||
| FQ228853.1 | - | 4 | 355 | |||

Bet1l
Genomic Map