siRNA Details
| Gene Symbol: Egln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019371.1 | NP_062244.1 | 1 | 374 | ||
| GenBank mRNA | U06713.1 | - | 1 | 374 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Egln3| Gene Symbol: Egln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019371.1 | NP_062244.1 | 1 | 374 | ||
| GenBank mRNA | U06713.1 | - | 1 | 374 | ||