siRNA Details
| Gene Symbol: Pdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271108.2 | NP_001258037.1 | 3 | 1306 | ||
| NM_019372.4 | NP_062245.2 | 2 | 1301 | |||
| XM_006237907.2 | XP_006237969.1 | 2 | 1421 | |||
| XM_006237908.2 | XP_006237970.1 | 2 | 1294 | |||
| XM_006237909.2 | XP_006237971.1 | 3 | 1385 | |||
| GenBank mRNA | AF062740.1 | - | 2 | 1283 | ||
| BC129095.1 | - | 2 | 1292 | 8369880 | ||

Pdp1
Genomic Map