siRNA Details
| Gene Symbol: Sept3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019375.1 | NP_062248.1 | 10 | 1020 | ||
| GenBank mRNA | AF111179.1 | - | 10 | 1020 | ||
| AF111180.1 | - | 8 | 1427 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sept3| Gene Symbol: Sept3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019375.1 | NP_062248.1 | 10 | 1020 | ||
| GenBank mRNA | AF111179.1 | - | 10 | 1020 | ||
| AF111180.1 | - | 8 | 1427 | |||