siRNA Details
| Gene Symbol: Nptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019380.1 | NP_062253.1 | 5 | 901 | ||
| XM_006243172.2 | XP_006243234.1 | 5 | 914 | |||
| XM_006243174.2 | XP_006243236.1 | 4 | 575 | |||
| XM_008766330.1 | XP_008764552.1 | 4 | 575 | |||
| GenBank mRNA | BC070947.1 | - | 4 | 431 | 7103647 | |
| X99337.1 | - | 4 | 604 | |||
| X99338.1 | - | 5 | 901 | |||

Nptn
Genomic Map