siRNA Details
| Gene Symbol: Nptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019380.1 | NP_062253.1 | 2 | 519 | ||
| XM_006243172.2 | XP_006243234.1 | 2 | 532 | |||
| GenBank mRNA | X99338.1 | - | 2 | 519 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nptn| Gene Symbol: Nptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019380.1 | NP_062253.1 | 2 | 519 | ||
| XM_006243172.2 | XP_006243234.1 | 2 | 532 | |||
| GenBank mRNA | X99338.1 | - | 2 | 519 | ||