siRNA Details
| Gene Symbol: Scaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019384.1 | NP_062257.1 | 4 | 715 | ||
| GenBank mRNA | U49056.1 | - | 4 | 715 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scaf1| Gene Symbol: Scaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019384.1 | NP_062257.1 | 4 | 715 | ||
| GenBank mRNA | U49056.1 | - | 4 | 715 | ||