siRNA Details
| Gene Symbol: Scaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019384.1 | NP_062257.1 | 4 | 1794 | ||
| XM_008759409.1 | XP_008757631.1 | 7 | 2757 | |||
| XM_008759410.1 | XP_008757632.1 | 7 | 3834 | |||
| XM_008759411.1 | XP_008757633.1 | 7 | 2704 | |||
| GenBank mRNA | U49056.1 | - | 4 | 1794 | ||

Scaf1
Genomic Map