siRNA Details
| Gene Symbol: Tgm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019386.2 | NP_062259.2 | 3, 4 | 498 | ||
| GenBank mRNA | AF106325.1 | - | 2, 3 | 449 | ||
| BC062062.1 | - | 3, 4 | 498 | 5599202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tgm2| Gene Symbol: Tgm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019386.2 | NP_062259.2 | 3, 4 | 498 | ||
| GenBank mRNA | AF106325.1 | - | 2, 3 | 449 | ||
| BC062062.1 | - | 3, 4 | 498 | 5599202 | ||