siRNA Details
| Gene Symbol: Mrpl37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004235.1 | NP_001004235.1 | 2 | 488 | ||
| GenBank mRNA | BC079328.1 | - | 2 | 488 | 7191681 | |
| FQ215540.1 | - | 2 | 501 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl37| Gene Symbol: Mrpl37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004235.1 | NP_001004235.1 | 2 | 488 | ||
| GenBank mRNA | BC079328.1 | - | 2 | 488 | 7191681 | |
| FQ215540.1 | - | 2 | 501 | |||