siRNA Details
| Gene Symbol: Notch3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020087.2 | NP_064472.2 | 13 | 2068 | ||
| GenBank mRNA | AF164486.2 | - | 13 | 2068 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Notch3| Gene Symbol: Notch3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020087.2 | NP_064472.2 | 13 | 2068 | ||
| GenBank mRNA | AF164486.2 | - | 13 | 2068 | ||