siRNA Details
| Gene Symbol: Srek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020092.1 | NP_064477.1 | 7 | 992 | ||
| XM_006231892.2 | XP_006231954.1 | 9 | 1511 | |||
| XM_006231894.2 | XP_006231956.1 | 8 | 5920 | |||
| GenBank mRNA | AF234765.1 | - | 7 | 992 | ||

Srek1
Genomic Map