siRNA Details
| Gene Symbol: Leprot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020099.2 | NP_064484.1 | 3 | 134 | ||
| GenBank mRNA | AF139209.1 | - | 3 | 161 | ||
| BC062003.1 | - | 3 | 134 | 5599858 | ||
| FQ225517.1 | - | 3 | 151 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Leprot| Gene Symbol: Leprot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020099.2 | NP_064484.1 | 3 | 134 | ||
| GenBank mRNA | AF139209.1 | - | 3 | 161 | ||
| BC062003.1 | - | 3 | 134 | 5599858 | ||
| FQ225517.1 | - | 3 | 151 | |||