siRNA Details
| Gene Symbol: Eif5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020075.1 | NP_064460.1 | 8 | 1229 | ||
| XM_006240608.1 | XP_006240670.1 | 10 | 1492 | |||
| XM_006240609.2 | XP_006240671.1 | 9 | 1772 | |||
| GenBank mRNA | BC062398.1 | - | 9 | 1417 | 5623373 | |
| FQ222051.1 | - | 6 | 1309 | |||
| L11651.1 | - | 8 | 1229 | |||
| Gene Symbol: RGD1564400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234307.2 | XP_006234369.1 | 1 | 618 | ||

Eif5
Genomic Map