siRNA Details
| Gene Symbol: Pth1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020073.2 | NP_064458.1 | 3 | 311 | ||
| XM_006243998.2 | XP_006244060.1 | 6 | 506 | |||
| XM_006243999.2 | XP_006244061.1 | 4 | 498 | |||
| XM_006244000.2 | XP_006244062.1 | 5 | 386 | |||
| XM_006244002.2 | XP_006244064.1 | 4 | 398 | |||
| XM_008766651.1 | XP_008764873.1 | 3 | 2410 | |||
| GenBank mRNA | L19475.1 | - | 3 | 311 | ||
| M77184.1 | - | 4 | 338 | |||
More Information
Set Membership: |
|

Pth1r
Genomic Map