siRNA Details
| Gene Symbol: Extl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020097.1 | NP_064482.1 | 1 | 1801 | ||
| XM_006252206.2 | XP_006252268.1 | 4 | 3107 | |||
| GenBank mRNA | AB033367.1 | - | 1 | 1801 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Extl3| Gene Symbol: Extl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020097.1 | NP_064482.1 | 1 | 1801 | ||
| XM_006252206.2 | XP_006252268.1 | 4 | 3107 | |||
| GenBank mRNA | AB033367.1 | - | 1 | 1801 | ||