siRNA Details
| Gene Symbol: Ramp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020100.2 | NP_064485.2 | 3 | 240 | ||
| GenBank mRNA | AB028935.1 | - | 2 | 41 | ||
| AB030944.1 | - | 3 | 219 | |||
| AB042889.1 | - | 3 | 238 | |||
| AF181552.2 | - | 3 | 240 | |||
| FQ214551.1 | - | 3 | 234 | |||
| FQ220330.1 | - | 3 | 221 | |||
| FQ220789.1 | - | 3 | 242 | |||
| FQ229563.1 | - | 3 | 259 | |||

Ramp3
Genomic Map