siRNA Details
| Gene Symbol: Cacna1i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020084.3 | NP_064469.3 | 6 | 1032 | ||
| GenBank mRNA | AF086827.2 | - | 1297 | |||
| AF290214.4 | - | 6 | 1032 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacna1i| Gene Symbol: Cacna1i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020084.3 | NP_064469.3 | 6 | 1032 | ||
| GenBank mRNA | AF086827.2 | - | 1297 | |||
| AF290214.4 | - | 6 | 1032 | |||