siRNA Details
Gene Symbol: Adam4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020305.1 | NP_064701.1 | 1 | 766 | ||
GenBank mRNA | BC107437.1 | - | 1 | 766 | 7132937 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adam4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020305.1 | NP_064701.1 | 1 | 766 | ||
GenBank mRNA | BC107437.1 | - | 1 | 766 | 7132937 |