siRNA Details
| Gene Symbol: Adam18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253341.2 | XP_006253403.1 | 14 | 1524 | ||
| XM_008771369.1 | XP_008769591.1 | 14 | 1524 | |||
| GenBank mRNA | Y11490.1 | - | 5 | 633 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam18| Gene Symbol: Adam18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253341.2 | XP_006253403.1 | 14 | 1524 | ||
| XM_008771369.1 | XP_008769591.1 | 14 | 1524 | |||
| GenBank mRNA | Y11490.1 | - | 5 | 633 | ||