siRNA Details
| Gene Symbol: Gstm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020540.1 | NP_065415.1 | 5 | 329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gstm3| Gene Symbol: Gstm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020540.1 | NP_065415.1 | 5 | 329 | ||