siRNA Details
Gene Symbol: Uts2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020537.1 | NP_065412.1 | 1 | 314 | ||
GenBank mRNA | AB012210.1 | - | 1 | 314 | ||
AB029611.1 | - | 1 | 314 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
