siRNA Details
| Gene Symbol: Tmem27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020976.1 | NP_066125.1 | 4 | 226 | ||
| GenBank mRNA | AF178086.1 | - | 4 | 226 | ||
| BC078838.1 | - | 4 | 283 | 7129929 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem27| Gene Symbol: Tmem27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020976.1 | NP_066125.1 | 4 | 226 | ||
| GenBank mRNA | AF178086.1 | - | 4 | 226 | ||
| BC078838.1 | - | 4 | 283 | 7129929 | ||