siRNA Details
| Gene Symbol: Apln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031612.3 | NP_113800.1 | 3 | 969 | ||
| GenBank mRNA | AF179679.1 | - | 3 | 929 | ||
| BC080843.1 | - | 3 | 955 | 7099981 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apln| Gene Symbol: Apln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031612.3 | NP_113800.1 | 3 | 969 | ||
| GenBank mRNA | AF179679.1 | - | 3 | 929 | ||
| BC080843.1 | - | 3 | 955 | 7099981 | ||