siRNA Details
Gene Symbol: Apln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031612.3 | NP_113800.1 | 3 | 1526 | ||
GenBank mRNA | BC080843.1 | - | 3 | 1512 | 7099981 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Apln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031612.3 | NP_113800.1 | 3 | 1526 | ||
GenBank mRNA | BC080843.1 | - | 3 | 1512 | 7099981 |