siRNA Details
| Gene Symbol: Tmod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031613.1 | NP_113801.1 | 3 | 377 | ||
| XM_006243407.2 | XP_006243469.1 | 4 | 837 | |||
| XM_006243408.2 | XP_006243470.1 | 4 | 4039 | |||
| XM_008766331.1 | XP_008764553.1 | 4 | 429 | |||
| GenBank mRNA | U59240.1 | - | 3 | 377 | ||

Tmod2
Genomic Map