siRNA Details
| Gene Symbol: Glrx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032614.2 | NP_116003.2 | 4 | 365 | ||
| XM_006230497.2 | XP_006230559.1 | 4 | 778 | |||
| GenBank mRNA | BC086381.1 | - | 4 | 365 | 7301390 | |
| FQ216152.1 | - | 1 | 15 | |||

Glrx3
Genomic Map