siRNA Details
| Gene Symbol: Glrx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032614.2 | NP_116003.2 | 5, 6 | 655 | ||
| XM_006230497.2 | XP_006230559.1 | 5, 6 | 1068 | |||
| GenBank mRNA | AF118651.1 | - | 5, 6 | 489 | ||
| BC086381.1 | - | 5, 6 | 655 | 7301390 | ||
| FQ216152.1 | - | 2, 3 | 305 | |||

Glrx3
Genomic Map