siRNA Details
| Gene Symbol: Zranb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031616.1 | NP_113804.1 | 1 | 63 | ||
| GenBank mRNA | AF013965.1 | - | 1 | 303 | ||
| AF013967.1 | - | 1 | 63 | |||
| BC087012.1 | - | 1 | 49 | 7189430 | ||
| FQ211774.1 | - | 1 | 89 | |||
| FQ211917.1 | - | 1 | 89 | |||

Zranb2
Genomic Map