siRNA Details
| Gene Symbol: Mest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009617.1 | NP_001009617.1 | 9, 10 | 1005 | ||
| GenBank mRNA | BC062800.1 | - | 9, 10 | 1005 | 6889877 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mest| Gene Symbol: Mest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009617.1 | NP_001009617.1 | 9, 10 | 1005 | ||
| GenBank mRNA | BC062800.1 | - | 9, 10 | 1005 | 6889877 | |