siRNA Details
| Gene Symbol: Sh2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031621.1 | NP_113809.1 | 2 | 963 | ||
| GenBank mRNA | U24655.1 | - | 2 | 963 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh2b3| Gene Symbol: Sh2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031621.1 | NP_113809.1 | 2 | 963 | ||
| GenBank mRNA | U24655.1 | - | 2 | 963 | ||