siRNA Details
| Gene Symbol: Sh2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031621.1 | NP_113809.1 | 2 | 1033 | ||
| GenBank mRNA | JX215366.1 | - | 1663 | |||
| U24653.1 | - | 5, 6 | 689 | |||
| U24654.1 | - | 4, 5 | 769 | |||
| U24655.1 | - | 2 | 1033 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh2b3| Gene Symbol: Sh2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031621.1 | NP_113809.1 | 2 | 1033 | ||
| GenBank mRNA | JX215366.1 | - | 1663 | |||
| U24653.1 | - | 5, 6 | 689 | |||
| U24654.1 | - | 4, 5 | 769 | |||
| U24655.1 | - | 2 | 1033 | |||