siRNA Details
| Gene Symbol: Nr1h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031626.1 | NP_113814.1 | 9 | 1300 | ||
| GenBank mRNA | BC157812.1 | - | 9 | 1279 | ||
| U14533.1 | - | 9 | 1300 | |||
| U20389.1 | - | 9 | 1244 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr1h2| Gene Symbol: Nr1h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031626.1 | NP_113814.1 | 9 | 1300 | ||
| GenBank mRNA | BC157812.1 | - | 9 | 1279 | ||
| U14533.1 | - | 9 | 1300 | |||
| U20389.1 | - | 9 | 1244 | |||