siRNA Details
| Gene Symbol: Ddx25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031630.2 | NP_113818.2 | 5 | 387 | ||
| GenBank mRNA | AF142629.2 | - | 5 | 387 | ||
| BC078791.1 | - | 5 | 369 | 7112991 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddx25| Gene Symbol: Ddx25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031630.2 | NP_113818.2 | 5 | 387 | ||
| GenBank mRNA | AF142629.2 | - | 5 | 387 | ||
| BC078791.1 | - | 5 | 369 | 7112991 | ||