siRNA Details
| Gene Symbol: Hpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053318.1 | NP_445770.1 | 10 | 1312 | ||
| GenBank mRNA | BC091137.1 | - | 10 | 1303 | 7326169 | |
| FQ210231.1 | - | 10 | 1314 | |||
| FQ211176.1 | - | 10 | 1314 | |||
| FQ218633.1 | - | 10 | 1311 | |||
| FQ218971.1 | - | 10 | 1302 | |||
| FQ218998.1 | - | 7 | 1081 | |||
| FQ219255.1 | - | 5 | 761 | |||
| FQ219309.1 | - | 10 | 1312 | |||

Hpx
Genomic Map