siRNA Details
| Gene Symbol: Casp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031632.1 | NP_113820.1 | 2 | 404 | ||
| GenBank mRNA | AF262319.1 | - | 2 | 404 | ||
| AF271996.1 | - | 2 | 371 | |||
| AF286006.1 | - | 2 | 404 | |||
| AF293333.1 | - | 2 | 404 | |||
| AF308469.1 | - | 2 | 371 | |||
| AF517560.1 | - | 2 | 404 | |||
| AY008275.1 | - | 2 | 371 | |||
| AY027667.1 | - | 2 | 380 | |||
| AY124461.1 | - | 2 | 404 | |||

Casp9
Genomic Map