siRNA Details
| Gene Symbol: Foxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031633.3 | NP_113821.2 | 8 | 1183 | ||
| XM_006237416.2 | XP_006237478.1 | 6 | 1126 | |||
| XM_006237417.2 | XP_006237479.1 | 6 | 1123 | |||
| XM_006237418.2 | XP_006237480.1 | 7 | 1150 | |||
| XM_006237419.2 | XP_006237481.1 | 5 | 1078 | |||
| XM_006237420.2 | XP_006237482.1 | 6 | 1102 | |||
| XM_006237421.2 | XP_006237483.1 | 7 | 1147 | |||
| GenBank mRNA | U83112.1 | - | 8 | 1183 | ||

Foxm1
Genomic Map