siRNA Details
| Gene Symbol: Pdcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031638.1 | NP_113826.1 | 1, 2 | 333 | ||
| GenBank mRNA | M80601.1 | - | 1, 2 | 115 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdcd2| Gene Symbol: Pdcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031638.1 | NP_113826.1 | 1, 2 | 333 | ||
| GenBank mRNA | M80601.1 | - | 1, 2 | 115 | ||