siRNA Details
| Gene Symbol: H2afz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022674.1 | NP_073165.1 | 4 | 278 | ||
| GenBank mRNA | BC060564.1 | - | 4 | 343 | 6922321 | |
| BC086348.1 | - | 4 | 323 | 7302603 | ||
| FQ213837.1 | - | 4 | 337 | |||
| FQ220829.1 | - | 4 | 339 | |||
| FQ221791.1 | - | 4 | 337 | |||
| FQ222470.1 | - | 4 | 337 | |||
| FQ224994.1 | - | 4 | 340 | |||
| FQ227672.1 | - | 4 | 339 | |||
| FQ229641.1 | - | 4 | 339 | |||
| FQ229746.1 | - | 4 | 339 | |||
| FQ232362.1 | - | 4 | 339 | |||
| FQ234498.1 | - | 4 | 339 | |||
| M37584.1 | - | 4 | 278 | |||
| X52316.1 | - | 4 | 249 | |||

H2afz
Genomic Map