siRNA Details
| Gene Symbol: Cacnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105733.1 | NP_001099203.1 | 9 | 992 | ||
| XM_006234173.2 | XP_006234235.1 | 10 | 885 | |||
| XM_008761773.1 | XP_008759995.1 | 9 | 4847 | |||
| GenBank mRNA | L02315.1 | - | 1031 | |||

Cacnb4
Genomic Map