siRNA Details
| Gene Symbol: Dlg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031639.1 | NP_113827.1 | 6 | 1165 | ||
| XM_006257080.2 | XP_006257142.1 | 6 | 1438 | |||
| XM_006257081.2 | XP_006257143.1 | 5 | 1383 | |||
| XM_006257082.2 | XP_006257144.1 | 6 | 1442 | |||
| XM_006257083.2 | XP_006257145.1 | 5 | 1386 | |||
| XM_008773267.1 | XP_008771489.1 | 6 | 1440 | |||
| GenBank mRNA | U50147.1 | - | 6 | 1165 | ||

Dlg3
Genomic Map